3-(Cyclohexylmethoxy)-benzoic acid 97%

Lieferant: Apollo Scientific

OR452065-1G OR452065-250MG
APOSOR452065-1GEA 568 EUR
APOSOR452065-1G APOSOR452065-250MG
3-(Cyclohexylmethoxy)-benzoic acid 97%
3-(Cyclohexylmethoxy)-benzoic acid
Formel: C₁₄H₁₈O₃
Molecular Weight: 234.291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11122824
CAS-Nummer: 158860-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR