3-Ethynyl-benzoic acid ethyl ester 97%
Lieferant: Apollo Scientific
OR452067-1G
APOSOR452067-1GEA
393
EUR
APOSOR452067-1G
3-Ethynyl-benzoic acid ethyl ester 97%
3-Ethynyl-benzoic acid ethyl ester
Formel:
C₁₁H₁₀O₂ Molecular Weight: 174,196 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13195274 CAS-Nummer: 178742-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...