((4-Iodophenyl)ethynyl)trimethylsilane 97%

Lieferant: Apollo Scientific

OR452068-1G OR452068-5G
APOSOR452068-1GEA 59.6 EUR
APOSOR452068-1G APOSOR452068-5G
((4-Iodophenyl)ethynyl)trimethylsilane 97%
((4-Iodophenyl)ethynyl)trimethylsilane
Formel: C₁₁H₁₃ISi
Lagertemperatur: Raumtemperatur
MDL: MFCD04974026
CAS-Nummer: 134856-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR