2-Bromo-5-(cyclopropyl)thiophene-3-carboxylic acid

Lieferant: Apollo Scientific

OR45207-1G OR45207-250MG OR45207-5G
APOSOR45207-1GEA 0 EUR
APOSOR45207-1G APOSOR45207-250MG APOSOR45207-5G
2-Bromo-5-(cyclopropyl)thiophene-3-carboxylic acid
2-Bromo-5-(cyclopropyl)thiophene-3-carboxylic acid
Formel: C₈H₇BrO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD30003636
CAS-Nummer: 2091719-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR