Boc-DL-norleucine 97%

Lieferant: Apollo Scientific

OR452085-1G OR452085-25G OR452085-5G
APOSOR452085-1GEA 0 EUR
APOSOR452085-1G APOSOR452085-25G APOSOR452085-5G
Boc-DL-norleucine 97%
Boc-DL-norleucine
Formel: C₁₁H₂₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01326593
CAS-Nummer: 125342-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR