Pyrrolidine-3-carboxylic acid methylamide hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
N-Methylpyrrolidine-3-carboxamide hydrochloride, N-Methyl-3-pyrrolidinecarboxamide hydrochloride
OR452107-1G
OR452107-250MG
APOSOR452107-1GEA
163
EUR
APOSOR452107-1G
APOSOR452107-250MG
Pyrrolidine-3-carboxylic acid methylamide hydrochloride 97%
Pyrrolidine-3-carboxylic acid methylamide hydrochloride
Formel:
C₆H₁₃ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD21605763 CAS-Nummer: 1361115-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...