5-(Cyclopropyl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR45211-1G OR45211-250MG OR45211-5G
APOSOR45211-1GEA 0 EUR
APOSOR45211-1G APOSOR45211-250MG APOSOR45211-5G
5-(Cyclopropyl)thiophene-2-carboxylic acid
5-(Cyclopropyl)thiophene-2-carboxylic acid
Formel: C₈H₈O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD06802416
CAS-Nummer: 29481-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR