Methyl 5-(cyclopropyl)thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR45212-1G OR45212-250MG OR45212-5G
APOSOR45212-1GEA 522 EUR
APOSOR45212-1G APOSOR45212-250MG APOSOR45212-5G
Methyl 5-(cyclopropyl)thiophene-2-carboxylate
Methyl 5-(cyclopropyl)thiophene-2-carboxylate
Formel: C₉H₁₀O₂S
Molecular Weight: 182.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32219468
CAS-Nummer: 1021432-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR