(1-Methylpiperidin-4-yl)acetic acid hydrochloride 97%

Lieferant: Apollo Scientific

OR452122-1G OR452122-500MG
APOSOR452122-1GEA 522 EUR
APOSOR452122-1G APOSOR452122-500MG
(1-Methylpiperidin-4-yl)acetic acid hydrochloride 97%
(1-Methylpiperidin-4-yl)acetic acid hydrochloride
Formel: C₈H₁₆ClNO₂
Molecular Weight: 193.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12196931
CAS-Nummer: 67686-06-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR