1-Boc-2-acetyl-piperidine 97%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 2-acetylpiperidine-1-carboxylate
OR452124-1G
APOSOR452124-1GEA
91.5
EUR
APOSOR452124-1G
1-Boc-2-acetyl-piperidine 97%
1-Boc-2-acetyl-piperidine
Formel:
C₁₂H₂₁NO₃ Molecular Weight: 227,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953375 CAS-Nummer: 201991-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...