3-(Cyclopropyl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR45213-1G OR45213-250MG
APOSOR45213-1GEA 0 EUR
APOSOR45213-1G APOSOR45213-250MG
3-(Cyclopropyl)thiophene-2-carboxylic acid
3-(Cyclopropyl)thiophene-2-carboxylic acid
Formel: C₈H₈O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD13195793
CAS-Nummer: 29488-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR