1-Bromo-4-(3-bromopropyl)benzene 97%
Lieferant: Apollo Scientific
OR452135-1G
APOSOR452135-1GEA
119
EUR
APOSOR452135-1G
1-Bromo-4-(3-bromopropyl)benzene 97%
1-Bromo-4-(3-bromopropyl)benzene
Formel:
C₉H₁₀Br₂ Molecular Weight: 277.984 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09744416 CAS-Nummer: 90562-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...