2-Amino-2-(3-methylphenyl)ethan-1-ol 97%
Lieferant: Apollo Scientific
OR452140-1G
OR452140-250MG
APOSOR452140-1GEA
0
EUR
APOSOR452140-1G
APOSOR452140-250MG
2-Amino-2-(3-methylphenyl)ethan-1-ol 97%
2-Amino-2-(3-methylphenyl)ethan-1-ol
Formel:
C₉H₁₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827183 CAS-Nummer: 1179634-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...