N-Boc-(S)-2-amino-1-butanol 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-tert-Butyl (1-hydroxybutan-2-yl)carbamate
OR452149-25G
APOSOR452149-25GEA
132
EUR
APOSOR452149-25G
N-Boc-(S)-2-amino-1-butanol 97%
N-Boc-(S)-2-amino-1-butanol
Formel:
C₉H₁₉NO₃ Molecular Weight: 189.252 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974275 CAS-Nummer: 150736-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...