(S)-N-Cbz-2-amino-1-phenylethylamine 97%

Lieferant: Apollo Scientific

OR452159-1G
APOSOR452159-1GEA 647 EUR
APOSOR452159-1G
(S)-N-Cbz-2-amino-1-phenylethylamine 97%
(S)-N-Cbz-2-amino-1-phenylethylamine
Formel: C₁₆H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09260584
CAS-Nummer: 130406-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR