4-(Cyclopropyl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR45216-1G OR45216-250MG OR45216-5G
APOSOR45216-1GEA 0 EUR
APOSOR45216-1G APOSOR45216-250MG APOSOR45216-5G
4-(Cyclopropyl)thiophene-2-carboxylic acid
4-(Cyclopropyl)thiophene-2-carboxylic acid
Formel: C₈H₈O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD22067668
CAS-Nummer: 1252903-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR