2-Amino-1-(4-methylphenyl)ethanol hydrochloride 97%

Lieferant: Apollo Scientific

OR452164-1G OR452164-500MG
APOSOR452164-1GEA 633 EUR
APOSOR452164-1G APOSOR452164-500MG
2-Amino-1-(4-methylphenyl)ethanol hydrochloride 97%
2-Amino-1-(4-methylphenyl)ethanol hydrochloride
Formel: C₉H₁₃NO·ClH
Molecular Weight: 187.667 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09028159
CAS-Nummer: 53360-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR