2-Amino-1-(4-methylphenyl)ethanol hydrochloride 97%
Lieferant: Apollo Scientific
OR452164-1G
OR452164-500MG
APOSOR452164-1GEA
633
EUR
APOSOR452164-1G
APOSOR452164-500MG
2-Amino-1-(4-methylphenyl)ethanol hydrochloride 97%
2-Amino-1-(4-methylphenyl)ethanol hydrochloride
Formel:
C₉H₁₃NO·ClH Molecular Weight: 187.667 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09028159 CAS-Nummer: 53360-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...