b-Amino-2-chlorobenzeneethanol 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-2-(2-chlorophenyl)ethanol

OR452169-1G
APOSOR452169-1GEA 190 EUR
APOSOR452169-1G
b-Amino-2-chlorobenzeneethanol 97%
b-Amino-2-chlorobenzeneethanol
Formel: C₈H₁₀ClNO
Molecular Weight: 171.624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750965
CAS-Nummer: 127428-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR