5-Bromo-4-(cyclopropyl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR45217-1G OR45217-250MG OR45217-5G
APOSOR45217-1GEA 522 EUR
APOSOR45217-1G APOSOR45217-250MG APOSOR45217-5G
5-Bromo-4-(cyclopropyl)thiophene-2-carboxylic acid
5-Bromo-4-(cyclopropyl)thiophene-2-carboxylic acid
Formel: C₈H₇BrO₂S
Molecular Weight: 247.109 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32219471
CAS-Nummer: 2231413-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR