5-Bromo-4-(cyclopropyl)thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
OR45217-1G
OR45217-250MG
OR45217-5G
APOSOR45217-1GEA
522
EUR
APOSOR45217-1G
APOSOR45217-250MG
APOSOR45217-5G
5-Bromo-4-(cyclopropyl)thiophene-2-carboxylic acid
5-Bromo-4-(cyclopropyl)thiophene-2-carboxylic acid
Formel:
C₈H₇BrO₂S Molecular Weight: 247.109 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD32219471 CAS-Nummer: 2231413-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...