b-Amino-4-nitrobenzenepropanol 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3-(4-nitrophenyl)propan-1-ol

OR452175-1G OR452175-250MG
APOSOR452175-1GEA 789 EUR
APOSOR452175-1G APOSOR452175-250MG
b-Amino-4-nitrobenzenepropanol 97%
b-Amino-4-nitrobenzenepropanol
Formel: C₉H₁₂N₂O₃
Molecular Weight: 196.203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17171137
CAS-Nummer: 174302-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR