Ethyl-2-iodbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Iodbenzoesäureethylester
OR4535-100G
APOSOR4535-100GEA
173
EUR
APOSOR4535-100G
Ethyl-2-iodbenzoat
Ethyl-2-iodbenzoat
Formel:
C₉H₉IO₂ Molecular Weight: 276,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051796 CAS-Nummer: 1829-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...