(E)-3-[4-(tert-Butyl)benzoyl]acrylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR4543-1G
APOSOR4543-1GEA 183 EUR
APOSOR4543-1G
(E)-3-[4-(tert-Butyl)benzoyl]acrylic acid 98%
(E)-3-[4-(tert-Butyl)benzoyl]acrylic acid
Formel: C₁₄H₁₆O₃
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00171556
CAS-Nummer: 97678-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR