2,4-Dimethoxypyrimidine-6-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dimethoxypyrimidine-4-carboxylic acid

OR4550-250MG
APOSOR4550-250MGEA 21.9 EUR
APOSOR4550-250MG
2,4-Dimethoxypyrimidine-6-carboxylic acid 95+%
2,4-Dimethoxypyrimidine-6-carboxylic acid
Formel: C₇H₈N₂O₄
Molecular Weight: 184,15 g/mol
Schmelzpunkt: 162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01568105
CAS-Nummer: 59864-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR