1-Acetyl-5-bromo-3-hydroxy-1H-indole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Acetyl-5-bromo-1H-indol-3-ol, 1-(5-Bromo-3-hydroxy-1H-inol-1-yl)ethan-1-one

OR4554-100MG
APOSOR4554-100MGEA 21.9 EUR
APOSOR4554-100MG
1-Acetyl-5-bromo-3-hydroxy-1H-indole
1-Acetyl-5-bromo-3-hydroxy-1H-indole
Formel: C₁₀H₈BrNO₂
Molecular Weight: 254,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00152012
CAS-Nummer: 114165-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR