(R)-(-)-3-Amino-1-Boc-piperidin
Lieferant: Apollo Scientific
|
Achtung
|
OR4589-25G
APOSOR4589-25GEA
0
EUR
APOSOR4589-25G
(R)-(-)-3-Amino-1-Boc-piperidin
(R)-(-)-3-Amino-1-Boc-piperidin
Chiral intermediate for pharmaceuticals and agrochemicals.
Formel:
C₁₀H₂₀N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094717 CAS-Nummer: 188111-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...