5-Bromisatosäureanhydrid 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
OR4597-5G
APOSOR4597-5GEA
21.9
EUR
APOSOR4597-5G
5-Bromisatosäureanhydrid 95+%
5-Bromisatosäureanhydrid
Formel:
C₈H₄BrNO₃ Molecular Weight: 242,03 g/mol Schmelzpunkt: 285…290 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016921 CAS-Nummer: 4692-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...