3-Brom-p-anisaldehyd 95%

Lieferant: Apollo Scientific

OR4599-100G
APOSOR4599-100GEA 97.8 EUR
APOSOR4599-100G
3-Brom-p-anisaldehyd 95%
3-Brom-p-anisaldehyd
Formel: C₈H₇BrO₂
Schmelzpunkt: 51…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016599
CAS-Nummer: 34841-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR