2-Aminopyrimidine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR46000-1G OR46000-250MG
APOSOR46000-1GEA 94.4 EUR
APOSOR46000-1G APOSOR46000-250MG
2-Aminopyrimidine-4-carboxylic acid
2-Aminopyrimidine-4-carboxylic acid
Formel: C₅H₅N₃O₂
Molecular Weight: 139.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07783936
CAS-Nummer: 2164-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR