4-Amino-5,6,7,8-tetrahydroisoquinoline
Lieferant: Apollo Scientific
OR460000-50MG
APOSOR460000-50MGEA
176
EUR
APOSOR460000-50MG
4-Amino-5,6,7,8-tetrahydroisoquinoline
4-Amino-5,6,7,8-tetrahydroisoquinoline
Formel:
C₉H₁₂N₂ Molecular Weight: 148,205 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09863911 CAS-Nummer: 130831-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...