rel-[(1R,3R)-3-(Aminomethyl)cyclopentyl]carbamic acid 1,1-dimethylethyl ester

Lieferant: Apollo Scientific
Achtung

OR460006-25MG
APOSOR460006-25MGEA 197 EUR
APOSOR460006-25MG
rel-[(1R,3R)-3-(Aminomethyl)cyclopentyl]carbamic acid 1,1-dimethylethyl ester
rel-[(1R,3R)-3-(Aminomethyl)cyclopentyl]carbamic acid 1,1-dimethylethyl ester
Formel: C₁₁H₂₂N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD24842666
CAS-Nummer: 862700-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR