(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-HoPro-OMe.HCl, H-D-Homopro-OMe·HCl

OR460007-5G
APOSOR460007-5GEA 75.4 EUR
APOSOR460007-5G
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride 98%
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride
Formel: C₇H₁₄ClNO₂
Molecular Weight: 179.644 g/mol
Schmelzpunkt: 159…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00237412
CAS-Nummer: 18650-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR