Tetrahydropyran-4-sulphonyl chloride
Lieferant: Apollo Scientific
Synonyme:
Tetrahydro-2H-pyran-4-sulphonyl chloride, 2H-Pyran-4-sulphonylchloride,tetrahydro-, oxane-4-sulphonyl chloride
OR460012-1G
APOSOR460012-1GEA
379
EUR
APOSOR460012-1G
Tetrahydropyran-4-sulphonyl chloride
Tetrahydropyran-4-sulphonyl chloride
Formel:
C₅H₉ClO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787616 CAS-Nummer: 338453-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...