4-(3,4-Methylenedioxyphenyl)-3-thiosemicarbazide

Lieferant: Apollo Scientific

OR460017-1G
APOSOR460017-1GEA 120 EUR
APOSOR460017-1G
4-(3,4-Methylenedioxyphenyl)-3-thiosemicarbazide
4-(3,4-Methylenedioxyphenyl)-3-thiosemicarbazide
Formel: C₈H₉N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00060504
CAS-Nummer: 206761-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR