8-Amino-3-bromoimidazo[1,2-a]pyrazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromoimidazo[1,2-a]pyrazin-8-amine
OR460018-250MG
APOSOR460018-250MGEA
183
EUR
APOSOR460018-250MG
8-Amino-3-bromoimidazo[1,2-a]pyrazine
8-Amino-3-bromoimidazo[1,2-a]pyrazine
Formel:
C₆H₅BrN₄ Molecular Weight: 213,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09265736 CAS-Nummer: 117718-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...