Cyclopentylmethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cyclopentanmethanol
OR46003-5G
APOSOR46003-5GEA
21.9
EUR
APOSOR46003-5G
Cyclopentylmethanol
Cyclopentylmethanol
Formel:
C₆H₁₂O Molecular Weight: 100,16 g/mol Siedepunkt: 162…163 °C Dichte: 0,926 g/cm³ (25 °C) Flammpunkt: 62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001384 CAS-Nummer: 3637-61-4 EINECS: 222-861-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...