4-Chlor-3-iodbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR460050-5G
APOSOR460050-5GEA
75.4
EUR
APOSOR460050-5G
4-Chlor-3-iodbenzonitril 98%
4-Chlor-3-iodbenzonitril
Formel:
C₇H₃ClIN Molecular Weight: 263,4629 g/mol Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025674 CAS-Nummer: 914106-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...