3-Bromoquinolin-6-yl acetate

Lieferant: Apollo Scientific
Achtung

OR46014-250MG
APOSOR46014-250MGEA 0 EUR
APOSOR46014-250MG
3-Bromoquinolin-6-yl acetate
3-Bromoquinolin-6-yl acetate
Formel: C₁₁H₈BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19687231
CAS-Nummer: 1022151-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR