1-(Methylsulphonyl)piperidine-4-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR46015-1G
OR46015-250MG
OR46015-100MG
OR46015-500MG
OR46015-2.5G
OR46015-5G
APOSOR46015-1GEA
0
EUR
APOSOR46015-1G
APOSOR46015-250MG
APOSOR46015-100MG
APOSOR46015-500MG
APOSOR46015-2.5G
APOSOR46015-5G
1-(Methylsulphonyl)piperidine-4-carbaldehyde
1-(Methylsulphonyl)piperidine-4-carbaldehyde
Formel:
C₇H₁₃NO₃S Schmelzpunkt: 97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11582937 CAS-Nummer: 241134-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...