7-Bromo[1,2,4]triazolo[1,5-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-[1,2,4]triazolo[1,5-a]pyridine
OR46017-500MG
APOSOR46017-500MGEA
408
EUR
APOSOR46017-500MG
7-Bromo[1,2,4]triazolo[1,5-a]pyridine
7-Bromo[1,2,4]triazolo[1,5-a]pyridine
Formel:
C₆H₄BrN₃ Molecular Weight: 198,0201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699200 CAS-Nummer: 1053655-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...