Resveratrol 99%

Lieferant: Apollo Scientific
Achtung

OR46018-100G OR46018-500G OR46018-1KG
APOSOR46018-100GEA 98.6 EUR
APOSOR46018-100G APOSOR46018-500G APOSOR46018-1KG
Resveratrol 99%
Resveratrol
Formel: C₁₄H₁₂O₃
Schmelzpunkt: 261…267 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00133799
CAS-Nummer: 501-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR