5-Formylpyridine-3-boronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

OR46032-1G OR46032-5G
APOSOR46032-1GEA 98.9 EUR
APOSOR46032-1G APOSOR46032-5G
5-Formylpyridine-3-boronic acid pinacol ester
5-Formylpyridine-3-boronic acid pinacol ester
Formel: C₁₂H₁₆BNO₃
Lagertemperatur: Gefrierschrank
MDL: MFCD04038759
CAS-Nummer: 848093-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR