4-Brom-3-chlorpyridin 98%

Lieferant: Apollo Scientific

OR46036-10G
APOSOR46036-10GEA 88.7 EUR
APOSOR46036-10G
4-Brom-3-chlorpyridin 98%
4-Bromo-3-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04112577
CAS-Nummer: 73583-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR