4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester

OR46046-100G
APOSOR46046-100GEA 65.4 EUR
APOSOR46046-100G
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 97%
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
Formel: C₁₆H₂₁NO₆
Molecular Weight: 323.35 g/mol
Schmelzpunkt: 98…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065564
CAS-Nummer: 7536-58-5
EINECS: 231-406-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR