3-Bromo-5-nitrophenol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-hydroxynitrobenzene

OR46064-10G OR46064-1G OR46064-5G
APOSOR46064-10GEA 292 EUR
APOSOR46064-10G APOSOR46064-1G APOSOR46064-5G
3-Bromo-5-nitrophenol 96%
3-Bromo-5-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218.005 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09965960
CAS-Nummer: 116632-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR