3,6-Dimethylsalicylaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-3,6-dimethylbenzaldehyde, 3,6-Dimethyl-2-hydroxybenzaldehyde
OR46065-250MG
APOSOR46065-250MGEA
33.5
EUR
APOSOR46065-250MG
3,6-Dimethylsalicylaldehyd
3,6-Dimethylsalicylaldehyd
Formel:
C₉H₁₀O₂ Molecular Weight: 150,1700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037328 CAS-Nummer: 1666-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...