6-Chloro-1H-pyrrolo[3,2-b]pyridine 97%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-4-azaindole

OR46087-1G
APOSOR46087-1GEA 58 EUR
APOSOR46087-1G
6-Chloro-1H-pyrrolo[3,2-b]pyridine 97%
6-Chloro-1H-pyrrolo[3,2-b]pyridine
Formel: C₇H₅ClN₂
Molecular Weight: 152,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10574982
CAS-Nummer: 1021339-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR