Tridecanol (mixture of isomers)
Lieferant: Apollo Scientific
|
Achtung
|
OR46094-10G
OR46094-1G
APOSOR46094-10GEA
0
EUR
APOSOR46094-10G
APOSOR46094-1G
Tridecanol (mixture of isomers)
Tridecanol (mixture of isomers)
Formel:
C₁₃H₂₈O Siedepunkt: 155…156 °C/15mmHg Schmelzpunkt: 29…34 °C Dichte: 0,822 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01778419 CAS-Nummer: 26248-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...