2,6-Dibromnaphthalin 99%
Lieferant: Apollo Scientific
|
Gefahr
|
OR46098-1G
OR46098-25G
OR46098-5G
APOSOR46098-1GEA
26.1
EUR
APOSOR46098-1G
APOSOR46098-25G
APOSOR46098-5G
2,6-Dibromnaphthalin 99%
2,6-Dibromonaphthalene
Formel:
C₁₀H₆Br₂ Molecular Weight: 285.97 g/mol Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01026462 CAS-Nummer: 13720-06-4 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...